Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014674.2 | NP_055489.1 | 12 | 3697 | ||
XM_011534271.2 | XP_011532573.1 | 12 | 3891 | |||
XM_011534272.2 | XP_011532574.1 | 13 | 3690 | |||
GenBank mRNA | D86967.1 | - | 12 | 3624 |