Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243776.1 | NP_001230705.1 | 12 | 1804 | ||
NM_001243777.1 | NP_001230706.1 | 10 | 1682 | |||
NM_014679.4 | NP_055494.2 | 11 | 1760 | |||
XM_006718945.2 | XP_006719008.1 | 10 | 1643 | |||
XM_006718946.2 | XP_006719009.1 | 9 | 1574 | |||
XM_017018592.1 | XP_016874081.1 | 12 | 2950 | |||
XM_017018593.1 | XP_016874082.1 | 9 | 1565 | |||
XM_017018594.1 | XP_016874083.1 | 8 | 1457 | |||
GenBank mRNA | AY239292.1 | - | 11 | 1661 | ||
BC001233.2 | - | 11 | 1743 | 3451715 | ||
BC031048.1 | - | 11 | 1671 | |||
D42054.1 | - | 10 | 1497 |