Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL14A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021110.3 | NP_066933.1 | 48 | 5940 | ||
XM_006716651.3 | XP_006716714.1 | 48 | 5803 | |||
XM_017013809.1 | XP_016869298.1 | 48 | 5729 | |||
GenBank mRNA | BC014640.1 | - | 27 | 2939 | ||
BC083495.1 | - | 14 | 1446 | |||
BC140893.1 | - | 47 | 5709 | 9021761 | ||
Y11711.1 | - | 2 | 393 |