Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 19 | 3205 | ||
XM_011537044.2 | XP_011535346.1 | 20 | 3307 | |||
XM_011537045.2 | XP_011535347.1 | 19 | 3117 | |||
GenBank mRNA | AL833497.1 | - | 6 | 2425 | ||
BC136775.1 | - | 19 | 3043 | 9020765 | ||
BC136776.1 | - | 19 | 3044 | 9020766 | ||
BC144407.1 | - | 20 | 3110 | 9052928 | ||
BX647116.1 | - | 19 | 3158 | |||
L40392.1 | - | 5 | 1289 |