Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 2 | 469 | ||
XM_005251587.1 | XP_005251644.1 | 2 | 440 | |||
XM_017015139.1 | XP_016870628.1 | 3 | 262 | |||
GenBank mRNA | AB058718.2 | - | 2 | 346 | ||
AK127218.1 | - | 2 | 458 | |||
AL834191.1 | - | 2 | 229 | |||
BC136771.1 | - | 2 | 443 | 9020761 | ||
BC136773.1 | - | 2 | 443 | 9020763 | ||
BC144404.1 | - | 2 | 443 | |||
CN390893.1 | - | 2 | 483 |