Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skida1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028317.2 | NP_082593.2 | 1 | 3171 | ||
GenBank mRNA | AK012843.1 | - | 1 | 718 | ||
AK034607.1 | - | 1 | 550 | |||
AK141728.1 | - | 1 | 1723 | |||
BC050860.1 | - | 2 | 2910 | 6331376 |