Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037324.2 | NP_001032401.1 | 16 | 1987 | ||
NM_001100120.1 | NP_001093590.1 | 17 | 2212 | |||
NM_001100121.1 | NP_001093591.1 | 17 | 2074 | |||
NM_014693.3 | NP_055508.3 | 17 | 2323 | |||
GenBank mRNA | AB011176.1 | - | 17 | 2069 | ||
AF192531.1 | - | 17 | 1979 | |||
AF428263.1 | - | 17 | 2251 | |||
AF428264.1 | - | 16 | 1892 | |||
AF521189.1 | - | 17 | 2188 | |||
AK293891.1 | - | 16 | 1917 | |||
AK295270.1 | - | 10 | 1124 | |||
AK296541.1 | - | 16 | 1970 | |||
AK309236.1 | - | 13 | 1880 | |||
AY359003.1 | - | 16 | 1943 | |||
BC142950.1 | - | 16 | 1987 | 8860334 |