Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032085.1 | NP_114474.1 | 45 | 3392 | ||
GenBank mRNA | AJ005395.1 | - | 7 | 592 | ||
BC087039.1 | - | 45 | 3392 | 7097081 | ||
FQ228741.1 | - | 3 | 115 | |||
X70369.1 | - | 10 | 808 |