Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032085.1 | NP_114474.1 | 51 | 4505 | ||
GenBank mRNA | AJ005395.1 | - | 13 | 1705 | ||
BC087039.1 | - | 51 | 4505 | 7097081 | ||
FQ214933.1 | - | 4 | 721 | |||
FQ215579.1 | - | 4 | 738 | |||
FQ215621.1 | - | 5 | 986 | |||
FQ221501.1 | - | 4 | 688 | |||
FQ228663.1 | - | 2 | 203 | |||
FQ228741.1 | - | 9 | 1230 | |||
FQ229745.1 | - | 5 | 935 | |||
FQ230038.1 | - | 4 | 686 | |||
X70369.1 | - | 16 | 1921 |