Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFCAB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032437.3 | NP_115813.2 | 8 | 1355 | ||
XM_005271274.3 | XP_005271331.1 | 8 | 1155 | |||
XM_006710976.3 | XP_006711039.1 | 8 | 1249 | |||
XM_006710977.1 | XP_006711040.1 | 8 | 1277 | |||
XM_011542301.2 | XP_011540603.1 | 8 | 1270 | |||
XM_017002548.1 | XP_016858037.1 | 8 | 1110 | |||
GenBank mRNA | AB058702.1 | - | 8 | 1118 | ||
AK092219.1 | - | 8 | 1266 | |||
AK292220.1 | - | 8 | 1314 | |||
AK315850.1 | - | 8 | 1131 | |||
BC015814.1 | - | 8 | 1059 | 3915942 |