Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053570.1 | NP_446022.1 | 8 | 1146 | ||
GenBank mRNA | BC088313.1 | - | 8 | 1146 | 7324723 |