Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053786.1 | NP_446238.1 | 7 | 654 | ||
GenBank mRNA | AF208499.1 | - | 3 | 218 | ||
BC082110.1 | - | 7 | 654 | 7135071 |