Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014700.3 | NP_055515.1 | 2 | 1067 | ||
XM_005255713.3 | XP_005255770.1 | 2 | 1102 | |||
XM_005255717.3 | XP_005255774.1 | 2 | 1113 | |||
XM_005255718.3 | XP_005255775.1 | 2 | 1113 | |||
XM_011522765.2 | XP_011521067.1 | 1 | 17 | |||
XM_017023907.1 | XP_016879396.1 | 2 | 1113 | |||
GenBank mRNA | AB014565.1 | - | 2 | 1115 | ||
AF395731.1 | - | 2 | 1115 | |||
BC051360.1 | - | 2 | 1066 | 6646253 |