Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168270.1 | NP_001161742.1 | 11, 12 | 1130 | ||
NM_133794.2 | NP_598555.2 | 15, 16 | 1476 | |||
GenBank mRNA | AK004814.1 | - | 15, 16 | 1410 | ||
AK030554.1 | - | 15, 16 | 1423 | |||
AK088049.1 | - | 15, 16 | 1423 | |||
AK145632.1 | - | 15, 16 | 1427 | |||
AK155759.1 | - | 15, 16 | 1425 | |||
AK165454.1 | - | 15, 16 | 1423 | |||
AK167782.1 | - | 15, 16 | 1423 | |||
AK170826.1 | - | 15, 16 | 1425 | |||
AK171293.1 | - | 15, 16 | 1425 | |||
BC023023.1 | - | 11, 12 | 1130 | 5362353 | ||
BC079854.1 | - | 15, 16 | 1410 | 5717082 |