Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFAP1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146337.2 | NP_001139809.1 | 16 | 2001 | ||
NM_001323062.1 | NP_001309991.1 | 15 | 1902 | |||
NM_152406.3 | NP_689619.1 | 16 | 2001 | |||
XM_011537558.1 | XP_011535860.1 | 12 | 1551 | |||
XM_017009036.1 | XP_016864525.1 | 16 | 1978 | |||
GenBank mRNA | AK074185.1 | - | 16 | 1944 | ||
AK094067.1 | - | 7 | 1031 | |||
BC125093.1 | - | 16 | 1920 | 40118258 | ||
BC125094.1 | - | 16 | 1920 | 40118263 |