Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014705.3 | NP_055520.3 | 40 | 4550 | ||
XM_006716188.2 | XP_006716251.1 | 41 | 4564 | |||
XM_006716189.2 | XP_006716252.1 | 41 | 4564 | |||
XM_017012819.1 | XP_016868308.1 | 41 | 4937 | |||
XM_017012820.1 | XP_016868309.1 | 40 | 4473 | |||
XM_017012821.1 | XP_016868310.1 | 41 | 4491 | |||
XM_017012822.1 | XP_016868311.1 | 41 | 5082 | |||
XM_017012823.1 | XP_016868312.1 | 41 | 4491 | |||
XM_017012824.1 | XP_016868313.1 | 40 | 4464 | |||
XM_017012825.1 | XP_016868314.1 | 41 | 4491 | |||
GenBank mRNA | AB018259.2 | - | 25 | 2663 | ||
AK294722.1 | - | 10 | 1007 | |||
AK294820.1 | - | 10 | 1270 | |||
AY233380.1 | - | 40 | 4281 | |||
BC117688.2 | - | 40 | 4281 | 40072000 | ||
BC117689.1 | - | 42 | 4416 | 40072006 |