Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320531.1 | NP_001307460.1 | 21, 22 | 2854 | ||
NM_183061.2 | NP_898884.1 | 22, 23 | 2998 | |||
NR_135297.1 | 16, 17 | 2057 | ||||
XM_006713603.2 | XP_006713666.1 | 22, 23 | 3033 | |||
XM_011512718.1 | XP_011511020.1 | 21, 22 | 2836 | |||
XM_011512720.1 | XP_011511022.1 | 22, 23 | 2998 | |||
XM_011512721.1 | XP_011511023.1 | 23, 24 | 3176 | |||
XM_011512725.2 | XP_011511027.1 | 12, 13 | 1547 | |||
XM_017006246.1 | XP_016861735.1 | 20, 21 | 2701 | |||
XM_017006247.1 | XP_016861736.1 | 21, 22 | 2812 | |||
GenBank mRNA | AK128084.1 | - | 22, 23 | 2998 | ||
BC044801.1 | - | 21, 22 | 2823 | 4827529 | ||
BC143774.1 | - | 15, 16 | 1902 |