Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194334.2 | NP_919315.2 | 13 | 5660 | ||
GenBank mRNA | AK004526.1 | - | 1 | 796 | ||
AK129273.1 | - | 13 | 5644 | |||
BC030847.1 | - | 3 | 3069 | |||
BC056467.1 | - | 6 | 3971 | |||
BC108420.1 | - | 8 | 4265 |