Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290659.1 | NP_001277588.1 | 25 | 4200 | ||
NM_198294.3 | NP_938036.2 | 26 | 4376 | |||
GenBank mRNA | AB093299.1 | - | 15 | 2469 | ||
AK004560.1 | - | 1 | 329 | |||
BC047437.1 | - | 5 | 566 | |||
BC079914.1 | - | 25 | 4179 | 30668628 | ||
BC141367.1 | - | 25 | 4190 | 9055980 |