Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290659.1 | NP_001277588.1 | 26 | 5633 | ||
NM_198294.3 | NP_938036.2 | 27 | 5809 | |||
GenBank mRNA | AB093299.1 | - | 16 | 3902 | ||
AK004560.1 | - | 2 | 1762 | |||
AK031162.1 | - | 1 | 845 | |||
AK037077.1 | - | 1 | 79 | |||
BC013225.1 | - | 1 | 804 | |||
BC047437.1 | - | 6 | 1999 | |||
BC050873.1 | - | 2 | 919 | |||
BC079914.1 | - | 26 | 5612 | 30668628 | ||
BC141367.1 | - | 26 | 5623 | 9055980 |