Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ly6g5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001934.1 | NP_001001934.1 | 2 | 95 | ||
XM_008772747.1 | XP_008770969.1 | 3 | 782 | |||
GenBank mRNA | HE864429.1 | - | 2 | 135 |
Gene Symbol: Csnk2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | HE864433.1 | - | 1752 | |||
HE864434.1 | - | 1752 |