Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014713.4 | NP_055528.1 | 7 | 1586 | ||
GenBank mRNA | AK129875.1 | - | 7 | 1236 | ||
AY359028.1 | - | 7 | 1203 | |||
BC000421.2 | - | 7 | 1163 | 2964450 | ||
BC003158.1 | - | 7 | 1226 | 2958870 | ||
BC093075.1 | - | 7 | 1186 | 30402963 | ||
CD679337.1 | - | 1 | 428 | |||
D14696.1 | - | 7 | 1224 |