Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001984.2 | NP_001001984.2 | 18 | 3613 | ||
GenBank mRNA | AK141779.1 | - | 4 | 854 | ||
AK173084.1 | - | 7 | 1431 | |||
BC036264.1 | - | 6 | 1312 | |||
BC037117.1 | - | 11 | 2162 | |||
BC057051.1 | - | 5 | 986 | |||
BC076576.1 | - | 18 | 3178 | 6840591 |