Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nol4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001986.2 | NP_001001986.2 | 8 | 5405 | ||
NM_001134300.2 | NP_001127772.1 | 11 | 6190 | |||
GenBank mRNA | AK147524.1 | - | 11 | 6193 | ||
AK164107.1 | - | 1 | 3810 | |||
BC075648.1 | - | 10 | 5600 | 30620653 | ||
BC079552.1 | - | 8 | 5348 | |||
BC094234.1 | - | 7 | 5256 |