Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002239.3 | NP_001002239.2 | 3 | 151 | ||
GenBank mRNA | AK002770.1 | - | 3 | 107 | ||
AK011132.1 | - | 3 | 107 | |||
AK011133.1 | - | 3 | 107 | |||
AK088443.1 | - | 3 | 109 | |||
AK137077.1 | - | 3 | 107 | |||
BC003896.1 | - | 2 | 103 | |||
BC054424.1 | - | 2 | 94 | 5011645 | ||
BC090401.1 | - | 2 | 77 | |||
BC090990.1 | - | 3 | 106 | 30297115 | ||
BC091759.1 | - | 3 | 140 | 4982640 | ||
BC092091.1 | - | 2 | 94 | 5252962 | ||
BC094640.1 | - | 2 | 74 | |||
BC106165.1 | - | 2 | 90 | 5007649 | ||
BY031981.1 | - | 3 | 152 |
Gene Symbol: Hk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | J05277.1 | - | 3703 |