Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrgprb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002283.1 | NP_001002283.1 | 61 |
Gene Symbol: LOC100361204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759464.1 | XP_008757686.1 | 1 | 78 |
Gene Symbol: LOC100912474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748862.2 | XP_003748910.1 | 1 | 78 |
Set Membership: |
G-protein coupled receptor |