Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Top1mt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002798.1 | NP_001002798.1 | 11 | 1421 | ||
XM_006241784.2 | XP_006241846.1 | 11 | 1621 |