Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002807.2 | NP_001002807.1 | 3 | 452 | ||
XM_008772745.1 | XP_008770967.1 | 3 | 453 | |||
GenBank mRNA | BC099823.1 | - | 3 | 404 | 7932557 | |
EU148064.1 | - | 3 | 215 | |||
FM067949.1 | - | 3 | 452 | |||
FM115459.1 | - | 1 | 74 | |||
FQ219968.1 | - | 3 | 442 | |||
FQ227136.1 | - | 3 | 238 | |||
FQ227567.1 | - | 3 | 229 | |||
FQ227951.1 | - | 3 | 229 | |||
FQ233892.1 | - | 3 | 229 |