Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002807.2 | NP_001002807.1 | 5 | 654 | ||
XM_008772745.1 | XP_008770967.1 | 5 | 655 | |||
GenBank mRNA | EU148064.1 | - | 5 | 417 | ||
FM115459.1 | - | 3 | 276 | |||
FQ219968.1 | - | 5 | 644 | |||
FQ227136.1 | - | 5 | 440 | |||
FQ227567.1 | - | 5 | 431 | |||
FQ227951.1 | - | 5 | 431 | |||
FQ233892.1 | - | 5 | 431 |