Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atf6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002809.3 | NP_001002809.2 | 17 | 1979 | ||
XM_006255983.1 | XP_006256045.1 | 17 | 2104 | |||
GenBank mRNA | BC098631.1 | - | 17 | 1970 | 7103893 |