Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014737.2 | NP_055552.1 | 12 | 2043 | ||
NM_170774.1 | NP_739580.1 | 11 | 1901 | |||
XM_005260895.3 | XP_005260952.1 | 12 | 2109 | |||
XM_011529410.1 | XP_011527712.1 | 12 | 2043 | |||
XM_011529411.1 | XP_011527713.1 | 12 | 2115 | |||
XM_011529412.2 | XP_011527714.1 | 11 | 1964 | |||
XM_017028149.1 | XP_016883638.1 | 11 | 1938 | |||
XM_017028150.1 | XP_016883639.1 | 13 | 2246 | |||
XM_017028151.1 | XP_016883640.1 | 13 | 2213 | |||
XM_017028152.1 | XP_016883641.1 | 13 | 2290 | |||
XM_017028153.1 | XP_016883642.1 | 11 | 1991 | |||
GenBank mRNA | AY154470.1 | - | 12 | 2043 | ||
AY154471.1 | - | 9 | 1817 | |||
AY154472.1 | - | 11 | 1901 | |||
BC117118.1 | - | 11 | 1852 | 40125669 | ||
BC117120.1 | - | 10 | 1880 | 40125671 | ||
CR627436.1 | - | 11 | 5251 | |||
D79990.1 | - | 12 | 2043 |