Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014740.3 | NP_055555.1 | 6, 7 | 816 | ||
XM_011525522.1 | XP_011523824.1 | 5, 6 | 709 | |||
GenBank mRNA | AK290608.1 | - | 6, 7 | 797 | ||
BC003662.2 | - | 6, 7 | 726 | 2988931 | ||
BC004386.1 | - | 6, 7 | 747 | 3619386 | ||
BC011151.1 | - | 6, 7 | 751 | 3452439 | ||
CR456750.1 | - | 6, 7 | 594 | |||
D21853.1 | - | 6, 7 | 808 | |||
X79538.1 | - | 6, 7 | 667 |