Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002835.1 | NP_001002835.1 | 7 | 617 | ||
XM_006240284.2 | XP_006240346.1 | 7 | 923 | |||
XM_006240285.2 | XP_006240347.1 | 7 | 923 | |||
XM_008764785.1 | XP_008763007.1 | 8 | 1023 | |||
XM_008764786.1 | XP_008763008.1 | 8 | 990 | |||
GenBank mRNA | FQ222878.1 | - | 7 | 874 |