Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002851.1 | NP_001002851.1 | 1, 2 | 204 | ||
GenBank mRNA | AY623793.1 | - | 1, 2 | 204 |