Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 5 | 591 | ||
XM_006234204.1 | XP_006234266.1 | 4 | 489 | |||
XM_006234205.2 | XP_006234267.1 | 3 | 2725 | |||
XM_008761838.1 | XP_008760060.1 | 3 | 903 | |||
XM_008761839.1 | XP_008760061.1 | 3 | 1189 | |||
GenBank mRNA | AB098072.1 | - | 5 | 591 |