Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tanc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002854.1 | NP_001002854.1 | 12 | 1920 | ||
XM_006234204.1 | XP_006234266.1 | 11 | 1839 | |||
XM_006234205.2 | XP_006234267.1 | 10 | 4075 | |||
XM_006234207.2 | XP_006234269.1 | 7 | 2037 | |||
XM_008761838.1 | XP_008760060.1 | 10 | 2253 | |||
XM_008761839.1 | XP_008760061.1 | 10 | 2539 | |||
XM_008761840.1 | XP_008760062.1 | 8 | 3171 | |||
GenBank mRNA | AB098072.1 | - | 12 | 1920 |