Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002894.2 | NP_001002894.2 | 10 | 2829 | ||
GenBank mRNA | AK014932.1 | - | 6 | 982 | ||
AY596199.1 | - | 9 | 2705 | |||
AY673647.1 | - | 10 | 2829 | |||
BC141199.1 | - | 10 | 2813 | 9055811 |