Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003668.2 | NP_001003668.2 | 4, 5 | 792 | ||
GenBank mRNA | BC139321.1 | - | 4, 5 | 792 | 8862343 | |
BC139322.1 | - | 4, 5 | 792 | 8862344 |