Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003671.1 | NP_001003671.1 | 1 | 863 | ||
GenBank mRNA | AY013768.1 | - | 1 | 863 |