Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267059.1 | NP_001253988.1 | 15 | 1669 | ||
NM_001267060.1 | NP_001253989.1 | 14 | 1630 | |||
NM_001267061.1 | NP_001253990.1 | 15 | 1540 | |||
NM_014748.3 | NP_055563.1 | 15 | 1705 | |||
NR_049782.1 | 15 | 1830 | ||||
NR_049783.1 | 16 | 1803 | ||||
NR_049784.1 | 16 | 1779 | ||||
NR_049785.1 | 14 | 1712 | ||||
NR_049786.1 | 15 | 1661 | ||||
NR_049787.1 | 13 | 1512 | ||||
NR_049788.1 | 12 | 1442 | ||||
XM_011533203.1 | XP_011531505.1 | 14 | 1479 | |||
XM_017005405.1 | XP_016860894.1 | 13 | 1649 | |||
GenBank mRNA | AK129689.1 | - | 15 | 1589 | ||
AK222543.1 | - | 15 | 1588 | |||
AK293464.1 | - | 15 | 1521 | |||
AK295278.1 | - | 13 | 1391 | |||
AK297054.1 | - | 15 | 1714 | |||
AK298620.1 | - | 15 | 1540 | |||
AK298856.1 | - | 15 | 1542 | |||
AK300144.1 | - | 15 | 1552 | |||
AK300641.1 | - | 13 | 1383 | |||
AK308487.1 | - | 14 | 1591 | |||
AK316373.1 | - | 15 | 1484 | |||
BC002524.1 | - | 15 | 1566 | 3140610 | ||
BC002610.1 | - | 15 | 1555 | 3143104 | ||
BC014620.2 | - | 15 | 1566 | 3010648 | ||
BC050590.1 | - | 15 | 1560 | 6044270 | ||
D31764.1 | - | 15 | 1679 |