Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003708.1 | NP_001003708.1 | 5 | 657 | ||
GenBank mRNA | AY373378.1 | - | 5 | 657 |