Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003912.1 | NP_001003912.1 | 40 | 4860 | ||
GenBank mRNA | AK081042.1 | - | 19 | 2823 | ||
AK220337.1 | - | 2154 | ||||
BC054078.1 | - | 7 | 1329 | |||
BC072663.1 | - | 39 | 5383 | |||
BC079565.1 | - | 40 | 4860 | 30536257 | ||
BC079915.1 | - | 40 | 5135 |