Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003913.2 | NP_001003913.1 | 14, 15 | 1908 | ||
NM_001171582.1 | NP_001165053.1 | 14, 15 | 1908 | |||
GenBank mRNA | AK191707.1 | - | 2, 3 | 171 | ||
AK197362.1 | - | 2, 3 | 172 | |||
AK204191.1 | - | 2, 3 | 172 | |||
BC052042.1 | - | 14, 15 | 1760 | |||
BC058968.1 | - | 14, 15 | 1800 | |||
BC079643.1 | - | 14, 15 | 1761 | 6416029 |