Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003913.2 | NP_001003913.1 | 12, 13 | 1704 | ||
NM_001171582.1 | NP_001165053.1 | 12, 13 | 1704 | |||
GenBank mRNA | BC052042.1 | - | 12, 13 | 1556 | ||
BC058968.1 | - | 12, 13 | 1596 | |||
BC079643.1 | - | 12, 13 | 1557 | 6416029 |