Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003918.2 | NP_001003918.2 | 19 | 2190 | ||
GenBank mRNA | AF548565.1 | - | 19 | 2133 | ||
BC022588.1 | - | 4 | 1006 | |||
BC046963.1 | - | 19 | 2128 | |||
BC100666.1 | - | 14 | 1496 |