Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003919.1 | NP_001003919.1 | 21 | 2205 | ||
GenBank mRNA | AK049419.1 | - | 21 | 2242 | ||
AK076357.1 | - | 22 | 2235 | |||
AK152868.1 | - | 21 | 2193 | |||
BC049929.1 | - | 9 | 719 | |||
BC079656.1 | - | 21 | 2205 | 6854039 |