Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003919.1 | NP_001003919.1 | 27 | 3507 | ||
GenBank mRNA | AK076357.1 | - | 26 | 4307 | ||
BC079656.1 | - | 27 | 3507 | 6854039 |