Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003948.2 | NP_001003948.2 | 3 | 1480 | ||
GenBank mRNA | AK035542.1 | - | 3 | 1355 | ||
AK083256.1 | - | 3 | 1401 | |||
AK150971.1 | - | 3 | 1419 | |||
AK151267.1 | - | 3 | 1449 | |||
BC054112.1 | - | 2 | 1113 | |||
BC080290.1 | - | 3 | 1426 | 6816865 |