Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013910.2 | NP_038938.1 | 11, 12 | 2298 | ||
GenBank mRNA | AF176524.2 | - | 4, 5 | 719 | ||
AK012952.1 | - | 1, 2 | 165 | |||
AK031626.1 | - | 11, 12 | 2264 | |||
AK129479.1 | - | 22, 23 | 3735 | |||
BC057622.1 | - | 11, 12 | 2293 | 6402440 | ||
BC065090.1 | - | 22, 23 | 3934 | 6851421 |