Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003964.1 | NP_001003964.1 | 9 | 879 | ||
XM_006256250.2 | XP_006256312.1 | 8 | 902 | |||
XM_006256251.2 | XP_006256313.1 | 1 | 350 |